Learn More
Thermco Products HotLog Digital Datalogger
Description
- This robust data logger is tailored for temperature monitoring in sterilization, pasteurization, and applications up to 140°C, making it ideal for food, pharmaceutical, and laboratory use.
- Crafted from food-safe AISI 316L stainless steel, it is waterproof and pressure-tight to IP68 standards up to 10 bar.
- Offers a temperature range from -40°C to 140°C with an accuracy of ±0.25°C and a resolution of 0.01°C.
- Features a 60,000-record memory and MultiLog capability for up to 500 recordings, with adjustable logging intervals from 1 sec to 24 hr.
- Automatic F0 and L value calculations support food safety compliance.
- User-replaceable 3.6 V LiSoCl2 battery provides up to 3 Years of operation or 5,000,000 measurements.
- Data management is supported via downloadable LOG-Connect software and a USB readout unit.
- Compact dimensions of 23 x 40 mm and a weight of 75 g make it ideal for quality control in production and sterilization processes.
Specifications
Specifications
| Type | Temperature Datalogger |
| Display Type | No Display |
| Alarm Types | None |
| Length (Metric) Probe | 50 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.