Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOX11 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056424 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056424 0.1 mg
NB10056424T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056424 Supplier Novus Biologicals Supplier No. NB10056424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOX11 Polyclonal specifically detects HOX11 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen HOX11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Alias homeo box 11 (T-cell lymphoma 3-associated breakpoint), homeo box-11 (T-cell leukemia-3 associated breakpoint, homologous to DrosophilaNotch), Homeobox protein Hox-11, HOX11T-cell leukemia, homeobox 1, Proto-oncogene TCL-3, T-cell leukemia homeobox 1, T-cell leukemia homeobox protein 1, T-cell leukemia/lymphoma protein 3, TCL3MGC163402
Gene Symbols TLX1
Host Species Rabbit
Immunogen A peptide sequence corresponding to acids amino acids 306-318 (LQNLQPWSDDSTK) of human HOX11 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3195
Test Specificity Cross reacts with Mouse, Human and Rat. Predicted to react with Bovine, Chicken, Chimpanzee, Dog and Xenopus.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.