Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP149835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149835 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149835 Supplier Novus Biologicals Supplier No. NBP149835
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HOXA1 Polyclonal specifically detects HOXA1 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen HOXA1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_005513.1
Gene Alias BSAS, homeo box A1, homeobox A1, Homeobox protein Hox-1F, homeobox protein Hox-A1, Hox 1.6-like protein, HOX A1 homeodomain protein, HOX1, HOX1Fhomeobox 1F, lab-like protein, MGC45232
Gene Symbols HOXA1
Host Species Goat
Immunogen Peptide with sequence C-QPATYQTSGN corresponding to internal region according to NP_705873.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3198
Test Specificity This antibody is expected to recognize reported isoforms a and b (NP_005513.1; NP_705873.2).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.