Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA9 Antibody (SR1583), Novus Biologicals™
SDP

Catalog No. NB260136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260136 100 μg
NB258616 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260136 Supplier Novus Biologicals Supplier No. NBP322360100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HOXA9 Monoclonal antibody specifically detects HOXA9 in Human samples. It is validated for Western Blot

Specifications

Antigen HOXA9
Applications Western Blot
Classification Monoclonal
Clone SR1583
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias ABD-B, homeo box A9, homeobox A9, Homeobox protein Hox-1G, homeobox protein Hox-A9, homeobox protein HOXA9, homeodomain protein HOXA9, HOX1, HOX1.7, HOX1GMGC1934
Host Species Rabbit
Immunogen A synthesized peptide derived from human HOXA9 (Uniprot #: P31269)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3205
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.