Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXA9 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $620.58
Specifications
Antigen | HOXA9 |
---|---|
Concentration | Lot-Specific |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2886
![]() ![]() |
Active Motif
39826 |
10 μL |
Each for $156.65
|
|
|||||
50-199-2885
![]() ![]() |
Active Motif
39825 |
100 μL |
Each for $620.58
|
|
|||||
Description
HOXA9 is the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development.Specifications
HOXA9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P31269 | |
3205 | |
This HOXA9 antibody was raised against a peptide within the N-terminal region of human HOXA9. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Hoxa9 |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
ABD-B; D6a9; homeo box A9; homeobox A9; homeobox A9-like; Homeobox protein Hox-1.7; Homeobox protein Hox-1G; Homeobox protein Hox-A9; homeodomain protein HOXA9; HOX1; HOX1.7; Hox-1.7; HOX1G; HOXA9; Hoxa-9; Hoxa9l; RGD1310001 | |
Hoxa9 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title