Learn More
Description
Specifically detects HOXB13 in Human samples, and it is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
| Antigen | HOXB13 |
| Applications | Western Blot, Immunoprecipitation, ChIP Assay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500, Chromatin Immunoprecipitation (ChIP) |
| Formulation | PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 |
| Gene Alias | homeo box B13, homeobox B13, homeobox protein Hox-B13, PSGD |
| Gene Symbols | HOXB13 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of human HOXB13. The exact sequence is proprietary. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
