Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXB4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | HOXB4 |
---|---|
Concentration | 0.28 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3799
![]() ![]() |
Proteintech Group Inc
222561AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
HOX homeobox genes play a role both in the early stem cell function as well as in later stages of hematopoietic differentiation. HOXB4, specifically, is a homeotic selector gene implicated in self-renewal of definitive HSCs. Expression of HOXB4 in primitive progenitors combined with culture on hematopoietic stroma induced a switch to the definitive HSC phenotype. Activation of WNT signaling in HSCs induced increased expression of HOXB4. Perturbations of HOX gene expression can cause leukemia.Specifications
HOXB4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P17483 | |
3214 | |
HOXB4 Fusion Protein Ag17761 | |
Primary | |
-20°C | |
HOXB4 |
0.28 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
homeobox B4, Homeobox protein Hox 2.6, Homeobox protein Hox 2F, Homeobox protein Hox B4, HOX 2.6, HOX2, HOX2F, HOXB4 | |
HOXB4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title