Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXB8 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB054982 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054982 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054982 Supplier Novus Biologicals Supplier No. NBP298722FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXB8 Polyclonal specifically detects HOXB8 in Human samples. It is validated for Western Blot.

Specifications

Antigen HOXB8
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias homeo box 2D, homeo box B8, homeobox B8, Homeobox protein Hox-2.4, Homeobox protein Hox-2D, homeobox protein Hox-B8, HOX2, HOX2DHox-2.4
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human HOXB8.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3218
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.