Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXC12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP168929
Description
HOXC12 Polyclonal specifically detects HOXC12 in Mouse samples. It is validated for Western Blot.Specifications
HOXC12 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
HOC3F, homeo box 3F, homeo box C12, homeobox C12, Homeobox protein Hox-3F, homeobox protein Hox-C12, HOX3FHOX3 | |
Rabbit | |
30 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q8K5B8 | |
HOXC12 | |
Synthetic peptides corresponding to Hoxc12 (homeobox C12) The peptide sequence was selected from the N terminal of Hoxc12. Peptide sequence PLVNIHTGDTFYFPNFRASGAQLPGLPSLSYPRRDNVCSLPWPSAEPCNG. | |
Affinity purified | |
RUO | |
3228 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction