Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXC12 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen HOXC12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP168929
SDP
View Documents
Novus Biologicals
NBP168929
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

HOXC12 Polyclonal specifically detects HOXC12 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

HOXC12
Polyclonal
Rabbit
Q8K5B8
3228
Synthetic peptides corresponding to Hoxc12 (homeobox C12) The peptide sequence was selected from the N terminal of Hoxc12. Peptide sequence PLVNIHTGDTFYFPNFRASGAQLPGLPSLSYPRRDNVCSLPWPSAEPCNG.
Primary
Western Blot
Unconjugated
RUO
HOC3F, homeo box 3F, homeo box C12, homeobox C12, Homeobox protein Hox-3F, homeobox protein Hox-C12, HOX3FHOX3
HOXC12
IgG
30 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.