Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXC8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152085 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152085 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152085 Supplier Novus Biologicals Supplier No. NBP152085
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HOXC8 Polyclonal specifically detects HOXC8 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen HOXC8
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_073149.1
Gene Alias homeo box C8, homeobox C8, Homeobox protein Hox-3A, homeobox protein Hox-C8, HOX3, Hox-3.1, mouse, homolog of, HOX3Ahomeo box 3A
Gene Symbols HOXC8
Host Species Goat
Immunogen Peptide with sequence C-SLYGAQQEAS corresponding to internal region according to NP_073149.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3224
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse and Rat.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.