Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXD13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179989
Description
HOXD13 Polyclonal specifically detects HOXD13 in Mouse samples. It is validated for Western Blot.Specifications
| HOXD13 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| BDSD, homeo box 4I, homeo box D13, homeobox D13, Homeobox protein Hox-4I, homeobox protein Hox-D13, HOX4IBDE, SPD | |
| Rabbit | |
| 36000 kDa | |
| 100 μL | |
| Primary | |
| Zebrafish: 77%. | |
| Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_032301 | |
| HOXD13 | |
| Synthetic peptide directed towards the C terminal of human Hoxd13. Peptide sequence VPTNEVPARAKEVSFYQGYTSPYQHVPGYIDMVSTFGSGEPRHEAYISME. | |
| Affinity purified | |
| RUO | |
| 3239 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction