Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXD13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | HOXD13 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
HOXD13 Polyclonal specifically detects HOXD13 in Mouse samples. It is validated for Western Blot.Specifications
| HOXD13 | |
| Polyclonal | |
| Rabbit | |
| NP_032301 | |
| 3239 | |
| Synthetic peptide directed towards the C terminal of human Hoxd13. Peptide sequence VPTNEVPARAKEVSFYQGYTSPYQHVPGYIDMVSTFGSGEPRHEAYISME. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| BDSD, homeo box 4I, homeo box D13, homeobox D13, Homeobox protein Hox-4I, homeobox protein Hox-D13, HOX4IBDE, SPD | |
| HOXD13 | |
| IgG | |
| 36000 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title