Learn More
Invitrogen™ HOXD13 Polyclonal Antibody

Description
Immunogen sequence: SFYQGYTSPYQ HVPGYIDMVS TFGSGEPRHE AYISMEGYQS WTLANGWNSQ VYCTKDQPQG SHFWKSSFPG Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
Specifications
Specifications
| Antigen | HOXD13 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HOXD13 |
| Gene Accession No. | P35453 |
| Gene Alias | BDE; BDSD; homeo box 4I; homeo box D13; homeobox D13; homeobox protein Hox-4.8; Homeobox protein Hox-4I; homeobox protein Hox-D13; Hox-4.8; HOX4I; Hoxd13; SPD; SPD1; spdh |
| Gene Symbols | HOXD13 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.