Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HP1 alpha Antibody (3A11F8) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252434 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252434 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252434 Supplier Novus Biologicals Supplier No. NBP252434
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HP1 alpha Monoclonal specifically detects HP1 alpha in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.

Specifications

Antigen HP1 alpha
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone 3A11F8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha
Gene Symbols CBX5
Host Species Mouse
Immunogen Purified recombinant fragment of human CBX5 (AA:1-191) expressed in E. Coli.
Molecular Weight of Antigen 22.2 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 23468
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.