Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HP1 alpha Antibody, Novus Biologicals™
SDP

Catalog No. NB1002518 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002518 0.1 mL
NB1002518SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002518 Supplier Novus Biologicals Supplier No. NB1002518
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HP1 alpha Polyclonal specifically detects HP1 alpha in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen HP1 alpha
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500, Flow (Intracellular) 1:500
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P45973
Gene Alias Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha
Gene Symbols CBX5
Host Species Rabbit
Immunogen A synthetic peptide containing an N-terminal region of human HP1 alpha (within residues 1-100). [Swiss-Prot P45973]
Molecular Weight of Antigen 20 kDa
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 23468
Test Specificity Human. Expected to react with mouse, Arabidopsis suecica, Rye and Triticum aestivum due to sequence homology.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.