Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HP1 Alpha, Beta, and Gamma Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB170462 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170462 100 μg
NB170463 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170462 Supplier Novus Biologicals Supplier No. NBP318651100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HP1 Alpha, Beta, and Gamma Polyclonal antibody specifically detects HP1 Alpha, Beta, and Gamma in Human samples. It is validated for Western Blot

Specifications

Antigen HP1 Alpha, Beta, and Gamma
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation Whole antiserum
Gene Accession No. P45973
Gene Alias Antigen p25, CBXM31HP1-BETA, chromobox homolog 1, chromobox homolog 1 (Drosophila HP1 beta), chromobox homolog 1 (HP1 beta homolog Drosophila ), chromobox homolog 3, chromobox homolog 3 (Drosophila HP1 gamma), chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 1, chromobox protein homolog 3, chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, Heterochromatin protein 1 homolog beta, Heterochromatin protein 1 homolog gamma, heterochromatin protein 1-beta, heterochromatin protein HP1 gamma, Heterochromatin protein p25, heterochromatin protein p25 beta, heterochromatin-like protein 1, HP1, HP1 alpha, HP1 alpha homolog, HP1 beta, HP1 beta homolog, HP1 gamma, HP1A, HP1-ALPHA, HP1-GAMMA, HP1Hs alpha, HP1Hs-alpha, HP1Hsbeta, HP1Hs-beta, MOD1, Modifier 1 protein, Modifier 2 protein, p25beta
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human HP1 Alpha, Beta, and Gamma (Heterochromatin protein 1 homolog beta), using the full length recombinant GST tagged protein. The antibody also recognizes the alpha and gamma isoforms.
Purification Method Whole Antiserum
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23468
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.