Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HP1BP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544637
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA544637 100 μL
1 options

Catalog No. PIPA544637

Supplier: Invitrogen™ PA544637

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SEESVSTVEE QENETPPATS SEAEQPKGEP ENEEKEENKS SEETKKDEKD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

HP1BP3 is the component of heterochromatin, may be involved in chromatin structure and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HP1BP3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HP1BP3
Gene Accession No. Q5SSJ5
Gene Alias heterochromatin protein 1 binding protein 3; heterochromatin protein 1, binding protein 3; heterochromatin protein 1-binding protein 3; Hp1bp3; HP1BP74; HP1-BP74; Protein HP1-BP74
Gene Symbols HP1BP3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HP1BP3 (aa 71-120).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50809
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.