Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPa2 Mouse anti-Human, Unconjugated, Clone: DHIC2-2B4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS204525UG
Description
Detects Alpha cells of human pancreatic islets using this mouse monoclonal Anti-HPa2, clone DHIC2-2B4, Cat. No. MABS2045, tested for use in Immunohistochemistry (Paraffin) and Flow Cytometry.
Specifications
HPa2 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Clone DHIC2-2B4 specifically detects alpha cells in human pancreatic tissue sections. | |
Stable for 1 year at 2-8°C from date of receipt. | |
Purified |
Immunohistochemistry (Paraffin), Flow Cytometry | |
DHIC2-2B4 | |
Unconjugated | |
Human pancreas alpha cells | |
Trypsin-dissociated human pancreatic islet cells containing low levels of exocrine and ductal cells. | |
25 μg | |
Signaling | |
NA | |
Human | |
Antibodies | |
IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction