Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPa2 Mouse anti-Human, Unconjugated, Clone: DHIC2-2B4, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | HPa2 |
---|---|
Clone | DHIC2-2B4 |
Concentration | Please refer to lot specific datasheet. |
Applications | Immunohistochemistry (Paraffin), Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS2045-25
|
MilliporeSigma
MABS204525UG |
25 μg |
Each for $192.35
|
|
|||||
MABS2045-100
|
MilliporeSigma
MABS2045100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Detects Alpha cells of human pancreatic islets using this mouse monoclonal Anti-HPa2, clone DHIC2-2B4, Cat. No. MABS2045, tested for use in Immunohistochemistry (Paraffin) and Flow Cytometry.Specifications
HPa2 | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Human pancreas alpha cells | |
Trypsin-dissociated human pancreatic islet cells containing low levels of exocrine and ductal cells. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
DHIC2-2B4 | |
Immunohistochemistry (Paraffin), Flow Cytometry | |
Unconjugated | |
Mouse | |
Signaling | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NA | |
IgG1 λ | |
Protein G purified | |
Clone DHIC2-2B4 specifically detects alpha cells in human pancreatic tissue sections. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title