Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPa3 Antibody (HIC3-2D12), Novus Biologicals™
SDP

Catalog No. NB173799 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173799 0.025 mg
NB173798 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB173799 Supplier Novus Biologicals Supplier No. NBP318520SS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HPa3 Monoclonal antibody specifically detects HPa3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen HPa3
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HIC3-2D12
Concentration 2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50
Formulation PBS
Host Species Mouse
Immunogen Human pancreatic islets enriched in non-beta endocine cells.
Purification Method Ion exchange chromatography
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°CC.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.