Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPa3 Antibody (HIC3-2D12), Novus Biologicals™

Mouse Monoclonal Antibody
$201.00 - $469.50
Specifications
Antigen | HPa3 |
---|---|
Clone | HIC3-2D12 |
Concentration | 2 mg/ml |
Dilution | Flow Cytometry, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50 |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
HPa3 Monoclonal antibody specifically detects HPa3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
HPa3 | |
2 mg/ml | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS | |
IgM | |
Ion exchange chromatography |
HIC3-2D12 | |
Flow Cytometry, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50 | |
Monoclonal | |
Mouse | |
Human | |
Human pancreatic islets enriched in non-beta endocine cells. | |
Primary | |
Store at 4°CC. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title