Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPCAL1 Mouse anti-Human, Unconjugated, Clone: 1E10, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | HPCAL1 |
---|---|
Clone | 1E10 |
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC1139-25
|
MilliporeSigma
MABC113925UG |
25 μg |
Each for $192.35
|
|
|||||
MABC1139-100
|
MilliporeSigma
MABC1139100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Detects Hippocalcin-like protein 1 using this mouse monoclonal Anti-HPCAL1, clone 1E10, Cat. No. MABC1139, tested for use in Western Blotting and Immunoprecipitation.Specifications
HPCAL1 | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Hippocalcin-like protein 1;Calcium-binding protein BDR-1;HLP2;Visinin-like protein 3;VILIP-3 | |
HPCAL1;BDR1 | |
IgG2b κ | |
Protein G purified | |
Clone 1E10 specifically detects Hippocalcin-like protein 1 (HPCAL1) in human cells. | |
∽22 kDa observed; 22.13 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
1E10 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Apoptosis & Cancer | |
Purified mouse monoclonal antibody IgG2b in PBS without azide. | |
NP_602293.1 | |
Purified native and denatured human HPCAL1. | |
Primary | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title