Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPCL Antibody, Novus Biologicals™
SDP

Catalog No. NBP131571 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131571 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131571 Supplier Novus Biologicals Supplier No. NBP131571

Rabbit Polyclonal Antibody

HPCL Polyclonal specifically detects HPCL in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HPCL
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias 1600020H07Rik, 2,2-HPCL, 2-hydroxyacyl-CoA lyase 1, 2-hydroxyphytanoyl-CoA lyase2-hydroxyphytanol-CoA lyase, EC 4.1, HPCL, PHYH2FLJ55041, Phytanoyl-CoA 2-hydroxylase 2, phytanoyl-CoA hydroxylase 2
Gene Symbols HACL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human HACL1. The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26061
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.