Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPD Antibody, Novus Biologicals™
SDP

Catalog No. NBP131593 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131593 Supplier Novus Biologicals Supplier No. NBP131593
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HPD Polyclonal specifically detects HPD in Human samples. It is validated for Western Blot.

Specifications

Antigen HPD
Applications Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal
Gene Alias 4HPPDEC 1.13.11.27,4-HPPD, 4-hydroxyphenylpyruvate dioxygenase, GLOD3, glyoxalase domain containing 3,4-hydroxyphenylpyruvic acid oxidase, HPPDase, PPD
Gene Symbols HPD
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HPD. The exact sequence is proprietary.
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3242
Test Specificity Tested in human. Predicted cross-reactivity based on sequence identity: Rat (89%). Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.