Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPd3 Mouse anti-Human, Unconjugated, Clone: DHIC5-4D9, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | HPd3 |
---|---|
Clone | DHIC5-4D9 |
Concentration | Please refer to lot specific datasheet. |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS2040-25
|
MilliporeSigma
MABS204025UG |
25 μg |
Each for $192.35
|
|
|||||
MABS2040-100
|
MilliporeSigma
MABS2040100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-HPd3, clone DHIC5-4D9, Cat. No. MABS2040, is a mouse monoclonal antibody that detects pancreatic and hepatic ductal cells and has been tested for use in Immunohistochemistry (Paraffin).Specifications
HPd3 | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Human pancreas and liver duct cells | |
Human pancreas enriched in duct cells. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
DHIC5-4D9 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Signaling | |
Purified mouse monoclonal antibody IgM in PBS with 0.05% sodium azide. | |
NA | |
IgM κ | |
Purified by ion-exchange chromatography | |
Clone DHIC5-4D9 detects human pancreatic and liver duct cells. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title