Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPi1 Antibody (HIC0-4F9), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB338550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338550 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338550 Supplier Novus Biologicals Supplier No. NBP118872AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPi1 Monoclonal antibody specifically detects HPi1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)

Specifications

Antigen HPi1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HIC0-4F9
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias human pancreatic endocrine islet cell
Host Species Mouse
Immunogen Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells.
Purification Method Tissue culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.