Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPi2 Mouse anti-Human, Unconjugated, Clone: HIC1-2B4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS199925UG
Description
Anti-HPi2, clone HIC1-2B4, Cat. No. MABS1999, mouse monoclonal antibody that detects human pancreatic enriched islet cells. and has been tested for use in Flow Cytometry, Immunofluorescence, and Immunohistochemistry (Paraffin).
Specifications
| HPi2 | |
| Monoclonal | |
| Please refer to lot specific datasheet. | |
| Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Clone HIC1-2B4 detects human pancreatic enriched islet cells. | |
| Stable for 1 year at 2-8°C from date of receipt. | |
| Purified |
| Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry | |
| HIC1-2B4 | |
| Unconjugated | |
| Human pancreatic enriched islet cells | |
| Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells. | |
| 25 μg | |
| Signaling | |
| NP_001013269.1 | |
| Human | |
| Antibodies | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction