Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPi2 Mouse anti-Human, Unconjugated, Clone: HIC1-2B4, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
| Antigen | HPi2 |
|---|---|
| Clone | HIC1-2B4 |
| Concentration | Please refer to lot specific datasheet. |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABS1999-25
|
MilliporeSigma
MABS199925UG |
25 μg |
Each for $192.35
|
|
|||||
|
MABS1999-100
|
MilliporeSigma
MABS1999100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-HPi2, clone HIC1-2B4, Cat. No. MABS1999, mouse monoclonal antibody that detects human pancreatic enriched islet cells. and has been tested for use in Flow Cytometry, Immunofluorescence, and Immunohistochemistry (Paraffin).Specifications
| HPi2 | |
| Please refer to lot specific datasheet. | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Human pancreatic enriched islet cells | |
| Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells. | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. | |
| Antibodies |
| HIC1-2B4 | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Signaling | |
| Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| NP_001013269.1 | |
| IgG2b κ | |
| Protein G purified | |
| Clone HIC1-2B4 detects human pancreatic enriched islet cells. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title