Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPR Monoclonal Antibody (C17)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA541595
This item is not returnable.
View return policy
Description
HPR Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a haptoglobin-related protein that binds hemoglobin as efficiently as haptoglobin. Unlike haptoglobin, plasma concentration of this protein is unaffected in patients with sickle cell anemia and extensive intravascular hemolysis, suggesting a difference in binding between haptoglobin-hemoglobin and haptoglobin-related protein-hemoglobin complexes to CD163, the hemoglobin scavenger receptor. This protein may also be a clinically important predictor of recurrence of breast cancer.
Specifications
HPR | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P00739 | |
HPR | |
Recombinant protein Haptoglobin Related Protein. The antigen corresponds to amino acid range 70-332 of the target protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
C17 | |
Unconjugated | |
HPR | |
A-259H10.2; Haptoglobin-related locus; haptoglobin-related protein; HP; HPR | |
Mouse | |
Protein A/G | |
RUO | |
3250 | |
-20°C | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction