Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HPR Monoclonal Antibody (C17)
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | HPR |
---|---|
Clone | C17 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541595
|
Invitrogen™
MA541595 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543768
|
Invitrogen™
MA543768 |
20 μL |
Each for $213.50
|
|
|||||
Description
HPR Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a haptoglobin-related protein that binds hemoglobin as efficiently as haptoglobin. Unlike haptoglobin, plasma concentration of this protein is unaffected in patients with sickle cell anemia and extensive intravascular hemolysis, suggesting a difference in binding between haptoglobin-hemoglobin and haptoglobin-related protein-hemoglobin complexes to CD163, the hemoglobin scavenger receptor. This protein may also be a clinically important predictor of recurrence of breast cancer.Specifications
HPR | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P00739 | |
3250 | |
Recombinant protein Haptoglobin Related Protein. The antigen corresponds to amino acid range 70-332 of the target protein. | |
Antibody |
C17 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
HPR | |
A-259H10.2; Haptoglobin-related locus; haptoglobin-related protein; HP; HPR | |
HPR | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title