Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPRT Antibody (2G8), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP17404A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP17404A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP17404A32 Supplier Novus Biologicals Supplier No. NBP174045AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPRT Monoclonal specifically detects HPRT in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HPRT
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2G8
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase
Gene Symbols HPRT1
Host Species Mouse
Immunogen Recombinant human HPRT (1-218aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3251.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.