Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPRT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525909
Change view
Click to view available options
Quantity:
400 μL
Catalog No. Quantity
PIPA525909 400 μL
1 options

Catalog No. PIPA525909

Supplier: Invitrogen™ PA525909

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with hamster, mouse, porcine, non-human primate and rat based on sequence homology.

The protein is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPRT1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HPRT1
Gene Accession No. P00492
Gene Alias C81579; HGPRT; HGPRTase; HPGRT; Hprt; HPRT 1; HPRT B; Hprt1; hprt1 protein; hypoxanthine guanine phosphoribosyl transferase; hypoxanthine guanine phosphoribosyl transferase 1; hypoxanthine phosphoribosyl transferase; hypoxanthine phosphoribosyltransferase 1; hypoxanthine-guanine phosphoribosyltransferase; hypoxanthine-guanine phosphoribosyltransferase 1; id:ibd1344; id:ibd5108; testicular tissue protein Li 89; wu:fc10g09; zgc:56221; zgc:86608
Gene Symbols HPRT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 65-94 amino acids from the N-terminal region of human HPRT1.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3251
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.