Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV11 E7 Antibody (711-13)
SDP

Catalog No. NB11059903 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11059903 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11059903 Supplier Novus Biologicals Supplier No. NB11059903

Mouse Monoclonal Antibody

HPV11 E7 Monoclonal specifically detects HPV11 E7 in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Human Papilloma Virus, type 11, oncoprotein E7
Applications Western Blot, ELISA
Classification Monoclonal
Clone 711-13, 711-45
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation PBS pH 7.2 with 0.09% Sodium Azide
Gene Alias Human papillomavirus E7 protein
Host Species Mouse
Immunogen human papilloma virus (HPV), type 11, oncoprotein E7 (711-13, 711-45)
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity human papilloma virus (HPV), type 11, oncoprotein E7 (711-13, 711-45)
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1, IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.