Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 Antibody (rHPV16L1/1058), Biotin, Novus Biologicals™
SDP

Catalog No. NB381609 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381609 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381609 Supplier Novus Biologicals Supplier No. NBP308318B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV16 Monoclonal antibody specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen HPV16
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rHPV16L1/1058
Conjugate Biotin
Formulation PBS
Gene Alias Human Papilloma Virus Type 16
Host Species Mouse
Immunogen Human HPV16, major capsid protein L1
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.