Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 E1+E4 Antibody (TVG 403), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB1002767R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002767R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002767R Supplier Novus Biologicals Supplier No. NB1002767R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV16 E1+E4 Monoclonal specifically detects HPV16 E1+E4 in Virus samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen HPV16 E1+E4
Applications CyTOF, ELISA, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone TVG 403
Conjugate DyLight 550
Dilution Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ATP dependent helicase E1, E1, E4, HPV 16 ATP dependent helicase E1, HPV 16 E1, HPV 16 E4, HPV 16 replication protein E1, HPV16 ATP dependent helicase E1, HPV16 E1, HPV16 E4, HPV16 replication protein E1, Human papilloma virus type 16, Human papilloma virus type 16 ATP dependent helicase E1, Human papilloma virus type 16 E1, Human papilloma virus type 16 E4, Human papilloma virus type 16 replication protein E1, Human papillomavirus type 16, Human papillomavirus type 16 ATP dependent helicase E1, Human papillomavirus type 16 E1, Human papillomavirus type 16 E4, Human papillomavirus type 16 replication protein E1, Replication protein, Replication protein E1
Host Species Mouse
Immunogen Antigen for hybridoma production was expressed as a beta galactosidase fusion protein using an expression system and was consequently cleaved to release the E1/E4 polypeptide.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity The epitope recognized by this antibody survived brief fixation in 5% formaldehyde, but not prolonged fixation.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.