Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 E2 Antibody (EL58), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB203067 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB203067 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB203067 Supplier Novus Biologicals Supplier No. NBP250427JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV16 E2 Monoclonal antibody specifically detects HPV16 E2 in Virus samples. It is validated for Western Blot

Specifications

Antigen HPV16 E2
Applications Western Blot
Classification Monoclonal
Clone EL58
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias HPV16 regulatory protein E2, Human Papilloma Virus 16 E2, Human papilloma virus 16 regulatory protein E2, Human papillomavirus type 16 E2, Human papillomavirus type 16 regulatory protein E2, Regulatory protein E2
Host Species Mouse
Immunogen Bacterial recombinant protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.