Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 E2 Antibody (TVG 261), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB384106 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384106 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384106 Supplier Novus Biologicals Supplier No. NBP308844DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV16 E2 Monoclonal antibody specifically detects HPV16 E2 in Virus samples. It is validated for Western Blot

Specifications

Antigen HPV16 E2
Applications Western Blot
Classification Monoclonal
Clone TVG 261
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias HPV16 regulatory protein E2, Human Papilloma Virus 16 E2, Human papilloma virus 16 regulatory protein E2, Human papillomavirus type 16 E2, Human papillomavirus type 16 regulatory protein E2, Regulatory protein E2
Host Species Mouse
Immunogen Raised against Vaccinia-E2 followed by intravenous injection of the maltose binding protein MBP-E2
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.