Learn More
Description
Specifications
Specifications
| Antigen | HPV16 E2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | TVG 261 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HPV16 regulatory protein E2, Human Papilloma Virus 16 E2, Human papilloma virus 16 regulatory protein E2, Human papillomavirus type 16 E2, Human papillomavirus type 16 regulatory protein E2, Regulatory protein E2 |
| Host Species | Mouse |
| Immunogen | Raised against Vaccinia-E2 followed by intravenous injection of the maltose binding protein MBP-E2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
