Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 E7 Antibody (3D6F1), Novus Biologicals™
SDP

Catalog No. NBP151548 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151548 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151548 Supplier Novus Biologicals Supplier No. NBP151548
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV16 E7 Monoclonal specifically detects HPV16 E7 in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Papillomavirus 16 E7
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3D6F1
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000
Gene Alias E7, HPV16 E7, HpV16gp2, Links Transforming protein human papillomavirus 16, Protein E7, Transforming protein, Transforming protein human papillomavirus 16
Host Species Mouse
Immunogen Purified full length E7 expressed in E. Coli.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Human papillomavirus type 16.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.