Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 E7 Antibody (716-332), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB10073151N Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073151N 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073151N Supplier Novus Biologicals Supplier No. NB10073151AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human Papilloma Virus Monoclonal specifically detects Human Papilloma Virus in Virus samples. It is validated for Sandwich ELISA, Western Blot.

Specifications

Antigen Human Papilloma Virus
Applications Sandwich ELISA, Western Blot
Classification Monoclonal
Clone 716-332
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias E7, E7 protein, HPV16 E7, HpV16gp2, Human Papilloma Virus Type 18 E7 protein, Human papillomavirus E7 protein, Links Transforming protein human papillomavirus 16, Protein E7, Transforming protein, Transforming protein human papillomavirus 16
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with HPV oncoprotein E7, type 16.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity HPV oncoprotein E7, types 16 and 18 (both monomer and dimer forms according WB)
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.