Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 E7 Antibody (TVG 701Y), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB1002768AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002768AG 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002768AG Supplier Novus Biologicals Supplier No. NB1002768AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Papillomavirus 16 E7 Monoclonal specifically detects Papillomavirus 16 E7 in Virus samples. It is validated for CyTOF, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen Papillomavirus 16 E7
Applications CyTOF, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone TVG 701Y
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias E7, HPV16 E7, HpV16gp2, Links Transforming protein human papillomavirus 16, Protein E7, Transforming protein, Transforming protein human papillomavirus 16
Host Species Mouse
Immunogen HPV16 E7 synthesised from S. pombe
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.