Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPV16/HPV18 E6 Mouse anti-Virus, Clone: HPV16/1295 + HPV18/1297, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30767520UG
Description
HPV16/HPV18 E6 Monoclonal specifically detects HPV16/HPV18 E6 in Virus samples. It is validated for ELISA.Specifications
HPV16/HPV18 E6 | |
Monoclonal | |
0.2 mg/ml | |
ELISA | |
E6, Human Papilloma Virus, Human papillomavirus type 16 E6, Human papillomavirus type 18 E6, Protein E6 | |
HPV18 E6-beta-galactosidase fusion protein | |
20 μg | |
Primary | |
Store at 4°C. | |
IgG1 κ |
ELISA | |
HPV16/1295 + HPV18/1297 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction