Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16/HPV18 E6 Mouse anti-Virus, Clone: HPV16/1295 + HPV18/1297, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen HPV16/HPV18 E6
Clone HPV16/1295 + HPV18/1297
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122824
SDP
View Documents
Novus Biologicals
NBP30767520UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

HPV16/HPV18 E6 Monoclonal specifically detects HPV16/HPV18 E6 in Virus samples. It is validated for ELISA.
Specifications

Specifications

HPV16/HPV18 E6
0.2 mg/ml
ELISA
Unconjugated
Mouse
Virus
E6, Human Papilloma Virus, Human papillomavirus type 16 E6, Human papillomavirus type 18 E6, Protein E6
IgG1 κ
Protein A or G purified
HPV16/1295 + HPV18/1297
ELISA
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
HPV18 E6-beta-galactosidase fusion protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.