Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 L1 Antibody (CamVir 1), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB003480 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003480 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003480 Supplier Novus Biologicals Supplier No. NB1002732DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV16 L1 Monoclonal specifically detects HPV16 L1 in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen HPV16 L1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Monoclonal
Clone CamVir 1
Conjugate DyLight 594
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HPV-16, HPV-16 capsid, HPV16 capsid protein, HPV16 L1, HPV16 major capsid protein L1, Human papillomavirus type 16 L1, Human papillomavirus type 16 major capsid protein L1, Major capsid protein, Major capsid protein L1
Host Species Mouse
Immunogen A beta galactosidase-L1 fusion protein purified by PAGE.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Cross reacts with HPV37. Reacts very strongly with biopsy specimins containing HPV16 or HPV33; very weak reactions were occasionally observed with biopsy specimens or smears containing HPV6 or HPV11.
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.