Learn More
Description
Specifications
Specifications
| Antigen | HPV16 L1 |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot |
| Classification | Monoclonal |
| Clone | CamVir 1 |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated |
| Formulation | PBS with No Preservative |
| Gene Alias | HPV-16, HPV-16 capsid, HPV16 capsid protein, HPV16 L1, HPV16 major capsid protein L1, Human papillomavirus type 16 L1, Human papillomavirus type 16 major capsid protein L1, Major capsid protein, Major capsid protein L1 |
| Host Species | Mouse |
| Immunogen | A beta galactosidase-L1 fusion protein purified by PAGE. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
