Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV16 L1 Antibody (CamVir 1), PE, Novus Biologicals™
SDP

Catalog No. NB339746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339746 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339746 Supplier Novus Biologicals Supplier No. NB1002732PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV16 L1 Monoclonal specifically detects HPV16 L1 in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen HPV16 L1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Monoclonal
Clone CamVir 1
Conjugate PE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation PBS
Gene Alias HPV-16, HPV-16 capsid, HPV16 capsid protein, HPV16 L1, HPV16 major capsid protein L1, Human papillomavirus type 16 L1, Human papillomavirus type 16 major capsid protein L1, Major capsid protein, Major capsid protein L1
Host Species Mouse
Immunogen A beta galactosidase-L1 fusion protein purified by PAGE.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.