Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV18 E6 Antibody (BF7), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB310333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310333 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310333 Supplier Novus Biologicals Supplier No. NB1002769MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV18 E6 Monoclonal antibody specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA, Radioimmune Assays

Specifications

Antigen HPV18 E6
Applications Western Blot, ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone BF7
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Radioimmunoassay
Formulation 50mM Sodium Borate
Gene Alias E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6
Host Species Mouse
Immunogen Gel-purified HPV18 E6-beta galactosidase fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.