Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV18 E6 Antibody (C1N1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB309108 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309108 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309108 Supplier Novus Biologicals Supplier No. NBP254807APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV18 E6 Monoclonal antibody specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen HPV18 E6
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone C1N1
Conjugate APC
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation PBS
Gene Alias E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6
Host Species Mouse
Immunogen HPV18E6-beta-galactosidase fusion protein synthesized in E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.