Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV18 E7 Antibody (718-15), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB359916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359916 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359916 Supplier Novus Biologicals Supplier No. NB11017215AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HPV18 E7 Monoclonal antibody specifically detects HPV18 E7 in Virus samples. It is validated for Western Blot, Immunoprecipitation, ELISA, KnockDown

Specifications

Antigen HPV18 E7
Applications Western Blot, Immunoprecipitation, ELISA, KnockDown
Classification Monoclonal
Clone 718-15
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunoprecipitation, Sandwich ELISA, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias E7 protein
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with HPV oncoprotein E7, type 18.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.